Indium(III) acetate, 99.99%, (trace metal basis), anhydrous, Thermo Scientific Chemicals
Indium(III) acetate, 99.99%, (trace metal basis), anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Indium(III) acetate, 99.99%, (trace metal basis), anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 436980500
Price (USD)/ Each
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS25114-58-3
IUPAC Nameindium(3+) triacetate
Molecular FormulaC6H9InO6
InChI KeyVBXWCGWXDOBUQZ-UHFFFAOYSA-K
SMILES[In+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration Complexometric35.3 to 43.3 % (In)
Total trace metal impurities=<100 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only