Gold(III) chloride, 99.99+%, (trace metal basis), Thermo Scientific Chemicals
Gold(III) chloride, 99.99+%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gold(III) chloride, 99.99+%, (trace metal basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 437080010
Price (USD)/ Each
206.00
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
206.00
Add to cart
Gold(III) chloride, 99.99+%, (trace metal basis), Thermo Scientific Chemicals
Catalog number437080010
Price (USD)/ Each
206.00
-
Add to cart
Chemical Identifiers
CAS13453-07-1
IUPAC Namegold hydrochloridyl
Molecular FormulaAuCl
InChI KeyBRSVJNYNWNMJKC-UHFFFAOYSA-N
SMILES[Cl].[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red to brown
Appearance (Form)Crystalline powder or crystals
Assay63.0 to 66.9 % (Au)
Total trace metal impurities=<100 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only