Thermo Scientific Chemicals

Rhodium(III) chloride hydrate, 99.95%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 437092500
250 mg, Glass bottle, Each
Thermo Scientific Chemicals

Rhodium(III) chloride hydrate, 99.95%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 437092500
250 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 437092500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
20765-98-4
IUPAC Name
rhodium(3+) trichloride
Molecular Formula
Cl3Rh
InChI Key
SONJTKJMTWTJCT-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Rh+3]
Appearance (Form)
Crystals
Assay
37.0 to 43.0 % (Rh)
Total trace metal impurities
=<500 ppm
Appearance (Color)
Red-brown to brown to black

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text