Thermo Scientific Chemicals

Iron, 99.99%, (trace metal basis), granules, Thermo Scientific Chemicals

Catalog number: 437110050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Iron, 99.99%, (trace metal basis), granules, Thermo Scientific Chemicals

Catalog number: 437110050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 437110050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7439-89-6
IUPAC Name
iron
Molecular Formula
Fe
InChI Key
XEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES
[Fe]
Appearance (Color)
Silver grey
Appearance (Form)
Granules
Particle size
0.5 to 1 mm
Total trace metal impurities
=<100 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text