Thermo Scientific Chemicals

Potassium hydroxide, 99.98%, (trace metal basis), pellets, Thermo Scientific Chemicals

Catalog number: 437135000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Potassium hydroxide, 99.98%, (trace metal basis), pellets, Thermo Scientific Chemicals

Catalog number: 437135000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 437135000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1310-58-3
IUPAC Name
potassium hydroxide
Molecular Formula
HKO
InChI Key
KWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES
[OH-].[K+]
Appearance (Color)
White
Appearance (Form)
Pellets
Titration with HCl
>=59.5 % (K)
Total trace metal impurities
=<200 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text