Gold, 99.99%, (trace metal basis), shot, Thermo Scientific Chemicals
Gold, 99.99%, (trace metal basis), shot, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gold, 99.99%, (trace metal basis), shot, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 437140010
Price (USD)/ Each
276.00
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
276.00
Add to cart
Gold, 99.99%, (trace metal basis), shot, Thermo Scientific Chemicals
Catalog number437140010
Price (USD)/ Each
276.00
-
Add to cart
Chemical Identifiers
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
Molecular Weight (g/mol)196.97
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Beads or pellets
Appearance (Color)Golden
Total trace metal impurities=<100 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only