Thermo Scientific Chemicals

Silicon dioxide, 99.998%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 437151000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Silicon dioxide, 99.998%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 437151000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 437151000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14808-60-7
IUPAC Name
silanedione
Molecular Formula
O2Si
InChI Key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES
O=[Si]=O
Total trace metal impurities
=<20 ppm
Appearance (Form)
Crystalline powder or crystals
Appearance (Color)
White
Assay
44.5 to 47.9 % (Si)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text