Hydrogen tetrachloroaurate hydrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Hydrogen tetrachloroaurate hydrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrogen tetrachloroaurate hydrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 437170010
Price (USD)/ Each
231.65
Online exclusive
257.00 
Save 25.35 (10%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
231.65
Online exclusive
257.00 
Save 25.35 (10%)
Add to cart
Hydrogen tetrachloroaurate hydrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Catalog number437170010
Price (USD)/ Each
231.65
Online exclusive
257.00 
Save 25.35 (10%)
-
Add to cart
Chemical Identifiers
CAS27988-77-8
IUPAC Namehydrogen tetrachlorogoldtris(ylium)
Molecular FormulaAuCl4H
InChI KeyRFWCEGCWRSNAGT-UHFFFAOYSA-K
SMILES[H+].Cl[Au+3](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Appearance (Color)Yellow to orange
Total trace metal impurities=<10 ppm
Assay49 to 51 % (Au)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only