Thermo Scientific Chemicals

Hydrogen tetrachloroaurate hydrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 437170050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Hydrogen tetrachloroaurate hydrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 437170050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 437170050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27988-77-8
IUPAC Name
hydrogen tetrachlorogoldtris(ylium)
Molecular Formula
AuCl4H
InChI Key
RFWCEGCWRSNAGT-UHFFFAOYSA-K
SMILES
[H+].Cl[Au+3](Cl)(Cl)Cl
Appearance (Form)
Crystals
Appearance (Color)
Yellow to orange
Total trace metal impurities
=<10 ppm
Assay
49 to 51 % (Au)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text