1x1 image pixel for data collection
L-Dihydroorotic acid, 99%
L-Dihydroorotic acid, 99%
L-Dihydroorotic acid, 99%
L-Dihydroorotic acid, 99%
Thermo Scientific Chemicals

L-Dihydroorotic acid, 99%

CAS: 5988-19-2 | C5H6N2O4 | 158.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4372400101 gGlass bottle
Catalog number 437240010
Price (USD)
150.65
Special offer
Online exclusive
Ends: 31-Dec-2025
177.00
Save 26.35 (15%)
Each
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
150.65
Special offer
Online exclusive
Ends: 31-Dec-2025
177.00
Save 26.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS5988-19-2
IUPAC Name(4S)-2,6-dioxo-1,3-diazinane-4-carboxylic acid
Molecular FormulaC5H6N2O4
InChI KeyUFIVEPVSAGBUSI-REOHCLBHSA-N
SMILESOC(=O)[C@@H]1CC(=O)NC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point253°C to 257°C
HPLC>=98.5 %
Appearance (Color)White to off-white
Infrared spectrumConforms
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link