5-Phenyl-2-furaldehyde, 96%, Thermo Scientific Chemicals
5-Phenyl-2-furaldehyde, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Phenyl-2-furaldehyde, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 437310050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name5-phenylfuran-2-carbaldehyde
Molecular FormulaC11H8O2
InChI KeyBMJHNNPEPBZULA-UHFFFAOYSA-N
SMILESO=CC1=CC=C(O1)C1=CC=CC=C1
Molecular Weight (g/mol)172.18
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to green to brown
Infrared spectrumConforms
GC>=95.0 %
Appearance (Form)Solid or liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only