3-Bromo-5-chloropyridine, 97%, Thermo Scientific Chemicals
3-Bromo-5-chloropyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromo-5-chloropyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 437400050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
IUPAC Name3-bromo-5-chloropyridine
Molecular FormulaC5H3BrClN
InChI KeyBELDOPUBSLPBCQ-UHFFFAOYSA-N
SMILESClC1=CN=CC(Br)=C1
Molecular Weight (g/mol)192.44
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<3.00 %
Appearance (Form)Crystalline powder
Melting point77.0°C to 81.0°C
Appearance (Color)White to brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only