Thermo Scientific Chemicals

3-Bromo-5-chloropyridine, 97%, Thermo Scientific Chemicals

Catalog number: 437400250
25 g, Each
Thermo Scientific Chemicals

3-Bromo-5-chloropyridine, 97%, Thermo Scientific Chemicals

Catalog number: 437400250
25 g, Each
Quantity
Catalog number: 437400250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
73583-39-8
IUPAC Name
3-bromo-5-chloropyridine
Molecular Formula
C5H3BrClN
InChI Key
BELDOPUBSLPBCQ-UHFFFAOYSA-N
SMILES
ClC1=CN=CC(Br)=C1
Infrared spectrum
Conforms
Water
=<3.00 %
Appearance (Form)
Crystalline powder
Melting point
77.0°C to 81.0°C
Appearance (Color)
White to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text