Invitrogen™

NA-Star™ Detection Microplates

Catalog number: 4374349
Have Questions?
Invitrogen™

NA-Star™ Detection Microplates

Catalog number: 4374349
Have Questions?
Catalog Number
4374349
Unit Size
10 plates
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
437434910 plates
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Solid white 96-well microplates, supplied with covers, optimized for use with the chemiluminescent NA-Star assay reagents. In addition, these microplates are ideal for use with other chemiluminescent assays that do not require a specific surface chemistry or coating. They do provide a high binding surface that can be coated with specific capture reagents for use with immunoassay⁄capture assay applications, if desired.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Barcode
No Barcode
Format
Standard
Detection Method
Colorimetric
Product Line
NA-Star NovaBright
For Use With (Equipment)
Luminometer (Microplate)
Plate Blocking
None
Quantity
10 plates
Shipping Condition
Room Temperature

Contents & Storage

Contains 10 plates with covers

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text