Thermo Scientific Chemicals

(S)-1-BOC-3-(Aminomethyl)piperidine, 97%, Thermo Scientific Chemicals

Catalog number: 437470050
5 g, Each
Thermo Scientific Chemicals

(S)-1-BOC-3-(Aminomethyl)piperidine, 97%, Thermo Scientific Chemicals

Catalog number: 437470050
5 g, Each
Quantity
Catalog number: 437470050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
140645-24-5
IUPAC Name
tert-butyl 3-(aminomethyl)piperidine-1-carboxylate
Molecular Formula
C11H22N2O2
InChI Key
WPWXYQIMXTUMJB-UHFFFAOYNA-N
SMILES
CC(C)(C)OC(=O)N1CCCC(CN)C1
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Specific optical rotation
-10° to -20° (20°C, 589 nm) (c=1, methanol)
Appearance (Form)
Liquid
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text