Applied Biosystems™

POROS™ MabCapture™ A Affinity Chromatography Resin

Catalog number: 4374735
Have Questions?
Applied Biosystems™

POROS™ MabCapture™ A Affinity Chromatography Resin

Catalog number: 4374735
Have Questions?
Catalog Number
4374735
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
250 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 L
Price (USD)
Availability
-
Quantity
-
Catalog Number
Unit Size
10 L
Price (USD)
Availability
-
Quantity
-
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
43747351 L
-
437473210 mL
-
437473050 mL
-
4374729250 mL
-
43747285 L
--
437473110 L
--
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Applied Biosystems™ POROS™ MabCapture™ A Media is a new design in Protein A chromatography resins technology, combining improvements made in bead and chemistry technology. These advances provide a monoclonal antibody purification resins that not only demonstrates the highest dynamic binding performance at low flow rates but also one that can maintain superior binding capacity at high linear velocities where other chromatography resins products cannot operate.

With POROS™ MabCapture™ affinity chromatography resins you can:

  • Obtain the highest dynamic binding capacity for monoclonal antibodies vs. conventional agarose or silica based Protein A medias
  • Maintain high dynamic binding capacity at flow rates over 700cm/hr
  • Design processes with greater flexibility (shorter bed heights, faster flow rates or both) due to the improvements made to the antibody binding efficiency
  • Clean and sanitize with standard sodium hydroxide agents utilized for other chromatography products
  • Recognize major increases to process productivity and reduce the downstream bottleneck

The development of humanized monoclonal antibodies has continued to fuel the growth of the biotherapeutic market. Addressing the therapeutic demand for monoclonal antibodies has lead to major improvements to the cell based expression systems that are used to manufacture these proteins. Consequently, these improvements have moved the processing bottleneck from upstream cell expression to the downstream purification process, as the purification operations struggle to keep up with the large quantities of antibodies that need to be manufactured.

Protein A chromatography, which is used as the primary capture chromatography step in most antibody process platforms, is generally regarded as the major process bottleneck due to the slow flow rates and long residence times required for the effective operation of conventional Protein A chromatography medias.The introduction of POROS™ MabCapture™ chromatography resins is aimed at cracking open the downstream bottleneck and providing the capacity, efficiency and flexibility needed to make improvements to the downstream purification platform resulting in major increases to process productivity and antibody manufacturing.

Pharmaceutical Grade Reagent. For Manufacturing and Laboratory Use Only.

Specifications

Form
Liquid Suspension
Product Line
POROS™
Quantity
1 L
Type
Bulk Resin
Stationary Phase
Protein A

Contents & Storage

Store refrigerated at 2°C to 8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text