Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The High Capacity cDNA Reverse Transcription Kit, available with or without RNase Inhibitor, contains all components necessary for the quantitative conversion of up to 2 μg of total RNA into single-stranded cDNA in a single 20 μL reaction. Both types of kit deliver extremely high-quality, single-stranded cDNA from 0.02–2 μg total RNA. Reactions can be scaled up to 100 μL to generate 10 μg of cDNA from a single reaction.
Format | Kit |
Reaction Speed | 120 min. |
Technique | Reverse Transcription |
Optimal Reaction Temperature | 37°C |
Reverse Transcriptase | MultiScribe |
Ribonuclease H Activity | Yes |
Shipping Condition | Dry Ice |
For Use With (Application) | RT-PCR |
Concentration | 50 U/μL |
Final Product Type | First-Strand cDNA |
No. of Reactions | 1,000 Reactions |
Reaction Format | Separate components |
Reagent Type | Reverse Transcription |
Size (Final Product) | Up to 7 kb |
Starting Material | RNA |
Includes | Kit with Rnase Inhibitor |
Unit Size | Each |
Format | Kit |
Reaction Speed | 120 min. |
Technique | Reverse Transcription |
Optimal Reaction Temperature | 37°C |
Reverse Transcriptase | MultiScribe |
Ribonuclease H Activity | Yes |
Shipping Condition | Dry Ice |
For Use With (Application) | RT-PCR |
Concentration | 50 U/μL |
Final Product Type | First-Strand cDNA |
No. of Reactions | 200 Reactions |
Reaction Format | Separate components |
Reagent Type | Reverse Transcription |
Size (Final Product) | 7 kb or Less |
Starting Material | RNA |
Includes | Kit only |
Unit Size | Each |
Format | Kit |
Reaction Speed | 120 min. |
Technique | Reverse Transcription |
Reverse Transcriptase | MultiScribe |
Ribonuclease H Activity | Yes |
Shipping Condition | Dry Ice |
For Use With (Application) | Real Time PCR (qPCR), RT-PCR |
Concentration | 50 U/μL |
Final Product Type | First-Strand cDNA |
No. of Reactions | 1,000 Reactions |
Reaction Format | Separate components |
Reagent Type | Reverse Transcription |
Size (Final Product) | 7 kb |
Starting Material | RNA |
Includes | Kit only |
Unit Size | Each |
Format | Kit |
Reaction Speed | 120 min. |
Technique | Reverse Transcription |
Reverse Transcriptase | MultiScribe |
Ribonuclease H Activity | Yes |
Shipping Condition | Dry Ice |
For Use With (Application) | Real Time PCR (qPCR), RT-PCR |
Concentration | 50 U/μL |
Final Product Type | First-Strand cDNA |
No. of Reactions | 200 Reactions |
Reaction Format | Separate components |
Reagent Type | Reverse Transcription |
Size (Final Product) | 7 kb |
Starting Material | RNA |
Includes | Kit with Rnase Inhibitor |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Includes | No. of Reactions | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
4374967 | Each | Kit with Rnase Inhibitor | 1,000 Reactions | 3,525.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4368814 | Each | Kit only | 200 Reactions | 464.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4368813 | Each | Kit only | 1,000 Reactions | 1,768.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4374966 | Each | Kit with Rnase Inhibitor | 200 Reactions | 760.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The High Capacity cDNA Reverse Transcription Kit with RNase Inhibitor includes the recombinant enzyme used to prevent degradation of RNA templates and lacks DNA endonuclease activity for better product yield.
In addition to their use in RT-PCR, this kit may be used in traditional PCR, microarrays, and for the generation of cDNA archives.
Features of the High Capacity cDNA Reverse Transcription Kit include:
• Linear target amplification for real-time PCR
• Higher yields and precision than other cDNA synthesis kits at a fraction of the cost
• 10-fold greater dynamic range than other kits
Extensively tested with a variety of templates
Quantitative first-strand synthesis of all RNA species is achieved with the use of the random primers. This kit has been extensively tested with a variety of RNA templates, including GC- and AU-rich targets and RNAs expressed at low levels. cRNA can also be efficiently generated from in vitro transcription of cDNA.