Thermo Scientific Chemicals

(1-BOC-Piperidin-4-yl)acetic acid, 97%, Thermo Scientific Chemicals

Catalog number: 437500010
1 g, Each
Thermo Scientific Chemicals

(1-BOC-Piperidin-4-yl)acetic acid, 97%, Thermo Scientific Chemicals

Catalog number: 437500010
1 g, Each
Quantity
Catalog number: 437500010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
157688-46-5
IUPAC Name
2-{1-[(tert-butoxy)carbonyl]piperidin-4-yl}acetate
Molecular Formula
C12H20NO4
InChI Key
ZXFLMSIMHISJFV-UHFFFAOYSA-M
SMILES
CC(C)(C)OC(=O)N1CCC(CC([O-])=O)CC1
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
Titration with NaOH
96 to 104 %
Appearance (Form)
Crystalline powder
Melting point
96°C to 102°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text