The Applied Biosystems™ MicroAmp™ Fast 48-Well Tray, specifically designed to snugly hold MicroAmp™ Fast Reaction Tubes with Cap (0.1 mL) or the MicroAmp™ Fast 8-Tube Strip (0.1 mL), is required for more efficient sample processing on the Applied Biosystems™ StepOne™ Real-Time PCR System.
• Engineered for efficient sample processing • Designed specifically for MicroAmp™ Fast Reaction Tubes with Cap (0.1 mL) and MicroAmp™ Fast 8-Tube Strip (0.1 mL) • Manufacturedto high specification, ensuring a quality product
Convenience and Performance The MicroAmp™ Fast 48-Well Tray allows you to organize your tubes while adding sample and mix. This handy tray reduces spillage and tube mix-ups by securing and ordering your tubes on the lab bench. When the samples are ready, simply put the tray of tubes directly on the block of the StepOne™ Real-Time PCR System. Not only does the tray eliminate the hassle of transferring the tubes, but it also provides the thermal contact necessary for thermal cycling.
Note: See user's manual or package insert for limited label license and trademark information.
Specifications
Certifications/Compliance
Certified DNA/RNase Free
Packaging
10 Trays
Product Line
MicroAmp
Product Type
48-Well Tray
For Use With (Equipment)
StepOne™
No. of Wells
48
For Use With (Application)
PCR
Plate Compatibility
48-Well Plates
Shipping Condition
Room Temperature
Contents & Storage
Contains 10 MicroAmp™ Fast 48-Well Trays. Store at room temperature.