4-Methyl-1-naphthaleneboronic acid, 96%, Thermo Scientific Chemicals
4-Methyl-1-naphthaleneboronic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methyl-1-naphthaleneboronic acid, 96%, Thermo Scientific Chemicals

Have Questions?
Quantity:
10 g
Catalog number 437530100
Price (USD)/ Each
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS103986-53-4
IUPAC Name(4-methylnaphthalen-1-yl)boronic acid
Molecular FormulaC11H11BO2
InChI KeyJHVQEUGNYSVSDH-UHFFFAOYSA-N
SMILESCC1=CC=C(B(O)O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
HPLC>=95.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only