5-Bromo-2-methylindole, 98%, Thermo Scientific Chemicals
5-Bromo-2-methylindole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromo-2-methylindole, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 437560010
Price (USD)/ Each
84.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
99.80 
Save 15.15 (15%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
84.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
99.80 
Save 15.15 (15%)
Add to cart
5-Bromo-2-methylindole, 98%, Thermo Scientific Chemicals
Catalog number437560010
Price (USD)/ Each
84.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
99.80 
Save 15.15 (15%)
-
Add to cart
Chemical Identifiers
CAS1075-34-9
IUPAC Name5-bromo-2-methyl-1H-indole
Molecular FormulaC9H8BrN
InChI KeyBJUZAZKEDCDGRW-UHFFFAOYSA-N
SMILESCC1=CC2=CC(Br)=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Yellow to pink to tan
Melting point103°C to 107°C
Infrared spectrumConforms
HPLC>=95.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only