Applied Biosystems™

Methyl Primer Express™ Software v1.0

Catalog number: 4376041
Have Questions?
Applied Biosystems™

Methyl Primer Express™ Software v1.0

Catalog number: 4376041
Have Questions?
Catalog Number
4376041
Unit Size
1 software
Price (USD)
Catalog NumberUnit SizePrice (USD)
43760411 software
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Methyl Primer Express™ Software v1.0 enables you to design high quality PCR primers for methylation mapping experiments. Simply cut and paste in your region of interest. The tool searches for CpG islands and simulates bisulfite modification of DNA in silico.

Methyl Primer Express™ Software v1.0 enables you to:

• Easily design robust primers for methylation studies
• Annotate the transcription start base and translation start codon
• Highlight target regions of interest and examine these boundaries in your primer design reports
• Predict CpG islands in your DNA sequence
• Simulate the bisulfite modification of DNA
• Create a “reference” sequence for downstream sequencing analysis

Modify CpG Search and Primer Design Criteria Easily
Flexible software enables you to easily modify the criteria you use to search for CpG islands in your sample DNA.

Create Information-Rich Reports
Create primer design reports containing your sample's initial DNA sequence, its bisulfite-modified sequence, a CpG density representation, the location of primers in the initial sequence, and both the primer sequence and its associated characteristics.

Part of a Complete Methylation Experiment Workflow
Applied Biosystems™ supplies all the tools you need for DNA methylation mapping experiments (see Figure 1 for details).

How to Order
Simply add Methyl Primer Express™ Software v1.0 to your cart and check out normally to receive a CD-ROM and printed Quick Reference Guide. Or download the software for free using the link below:

Download Methyl Primer Express™ Software v1.0

For Research Use Only. Not for use in diagnostics procedures.

Specifications

Product Line
Methyl Primer Express™
Software Type
Epigenetics, Primer Design
Data Entry
Genomic DNA sequence data
Data Outputs
CpG Site Reports, Bisulfite Sequencing Primer (BSP) Reports
For Use With (Equipment)
310 Genetic Analyzer, 3730 DNA Analyzer, 3500 Genetic Analyzer
License
Static
Operating System
Windows XP (Service Pack 2), Windows 2000 (Service Pack 4)
Product Type
Methyl Primer Express Software V1.0
Software Category
Software
System Requirements
Computer: Core 2 Duo E8400/3 GHz; 2 GB RAM

Contents & Storage

Contains one software CD and quick reference guide.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text