Informational:Informational:This product will be discontinued in June 2024. We recommend the QuantStudio qPCR family as an alternative.
Applied Biosystems™

StepOne™ Real-Time PCR System, desktop

Catalog number: 4376374
Have Questions?
Applied Biosystems™

StepOne™ Real-Time PCR System, desktop

Catalog number: 4376374
Have Questions?
Catalog Number
4376374
Unit Size
1 instrument
Price (USD)
Catalog NumberUnit SizePrice (USD)
43763741 instrumentRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The StepOne™ Real-Time PCR System is a 48-well, low-throughput Real-Time PCR instrument perfect for both first-time and experienced users. The StepOne™ Real-Time PCR System can be setup in a variety of configurations and comes ready to use, out of the box, with intuitive data analysis and instrument control software. Utilizing robust LED based 3-color optical recording, the StepOne™ Real-Time PCR System is designed to deliver precise, quantitative Real-Time PCR results for a variety of genomic research applications. Includes a tower computer.

Features of the The StepOne™ Real-Time PCR System include:

• Advanced software and instrumentation for performing a wide array of genomic assays.
• Sensitive 3-color optical LED recording system
• Intuitive and robust software perfect for both first-time and advanced users
• Simple and flexible instrument set-up and usage

StepOne™ Software
The StepOne™ Software included with the StepOne™ Real-Time PCR System runs on Windows™ XP, Window Vista™, and Windows™ 7 operating systems and provides instrument control, data collection, and data analysis capabilities. This latest version includes capabilities to collect melt curve data for High Resolution Melt (HRM) applications and the option to export in Real-Time PCR Data Markup Language (RDML) for compatibility with MIQE guidelines.

Rapid and Simple Assay Set-Up
This remarkably simple Real-Time PCR system is designed with an LCD touchscreen and a user-friendly interface that brings the power of genetic studies to researchers new to Real-Time PCR (Fig 1). The intuitive software and protocol wizards help guide new users through their Real-Time PCR experiments. The StepOne™ Real-Time PCR System also includes a quick-start setup so that you start a run immediately and enter plate information at a later time.

Sensitive LED-Based 3-Color Fluorescence Reading
The StepOne™ Real-Time PCR System utilizes a long-life LED-based optical system that can record fluorescence from FAM™/SYBR™ Green, VIC™/JOE™, and ROX™ dyes. This cost-effective, 3-color, 48-well system delivers precise, quantitative Real-Time PCR results and saves data from all filters in every run without depending on a computer or plate setup. It can discriminate between 2 populations of 5,000 and 10,000 template copies of a TaqMan™ assay with 99.7% confidence.

Compatible with Many Genomic Analysis Techniques
Perform a variety of standard and demanding genetic analysis research techniques on one instrument using the user friendly StepOne™ Real-Time PCR System and software. The StepOne™ System supports many Real-Time PCR applications, including the following:

• SNP Genotyping
• Gene Expression Analysis
• MicroRNA Expression
• Translocation Analysis
• Gene Detection
• Viral Load Analysis

The included StepOne™ Software supports a variety of analysis methods, including the following:

• Standard Curve (absolute quantitation)
• Relative Standard Curve
• Comparative Ct (ΔΔCt) (relative quantitation)
• Genotyping and Presence/Absence
• Melt Curve Analysis
High Resolution Melting (additional software required)

Versatile Instrument Configuration Options
The ultra-compact footprint of a StepOne™ System can be installed in multiple distinct configurations, providing unmatched flexibility and convenience that can allow a fit in any laboratory (Fig 2). The StepOne™ Real-Time PCR System can be used via a PC, with a PC connected to a Local Area Network (LAN), or as a stand-alone instrument (PC-free). Each StepOne™ Real-Time PCR System is factory-calibrated for optical and thermal accuracy, so simply remarkable Real-Time PCR results can be obtained right out of the box.

Real-Time Data Monitoring, Dissemination, and Storage
The system measures amplification as it occurs, allowing you to monitor the progress of your experiment cycle by cycle, either on the machine or remotely. Your data is stored on the instrument itself, and can be viewed and stored automatically via remote access or transferred via email or a USB flash drive. Data can also be conveniently exported as PowerPoint™, Excel™, and graphical image files.

Expanded Gene Expression Capabilities
The advanced software provided with the StepOne™ Real-Time PCR System now includes the powerful Gene Expression Study Package. This software package allows for greater flexibility and accuracy in your gene expression assays through:

• Analysis of an unlimited number of plates in one study
• Sorting of data by biological or technical replicate group
• Use of multiple endogenous controls
• Assay efficiency correction

High Resolution Melting Capability
Now with High Resolution Melt Software v3.0 you can perform post-PCR sequence variation analysis with the StepOne™ Real-Time PCR System. This separately purchased software simplifies set-up by maintaining assay specific settings and accepts pasted plate layout information directly from Excel. HRM Software v3.0 also has an improved clustering algorithm for increased sensitivity and accuracy, and the ability to conduct separate analyses of multiple assays run on a single plate.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
48-well Plate, 0.1 ml Tubes
Thermal Accuracy
± 0.25°C (35°C to 95°C) of Display Temperature
Sample Ramp Rate
Fast Mode: ± 2.2°C⁄s, Standard mode: ± 1.6°C⁄s
Sensitivity
1 Copy
Temperature Range (Metric)
4 to 100°C
Reaction Volume Range
10-30 μl (Standard Curve Experiments: 40 μl in Standard Mode is Validated)
Passive Reference Dye
ROX (Separate Tube), No ROX, ROX (Pre-mixed)
Dynamic Range
Linear Dynamic Range greater than 9 Log Units (Detection)
Calibrated Dye
VIC™, SYBR™ Green I, JOE™, FAM™, ROX™
High-throughput Compatibility
Multiplexing
For Use With (Equipment)
StepOne™
Thermal Uniformity
± 0.50°C (35°C to 95°C) of Setpoint/Display Temperature
Reaction Speed
Fast, Standard
Detection Method
SYBR, Primer-Probe Detection
Run Time
<2 hrs⁄run (Standard Mode), <40 min⁄run (Fast Mode)
Optics
LED, 3 Emission Filters, Photodiode
Thermal Cycling System
Peltier-based System
Dimensions
24.6 cm/9.7 in.(W) x 42.7 cm/16.8 in.(D) x 51.2 cm/20.2 in.(H)
Capacity
48 x 0.1 ml Tubes, 1 x 48-well Plate
External Computer
Minitower
Product Line
StepOne™
Peak Block Ramp Rate
4.6°C⁄s
Display Type
LCD Touchscreen
Green Features
Energy Efficient, Fewer Resources used and Less Waste
Weight
23.6 kg

Contents & Storage

Desktop

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text