2-Amino-3-methoxybenzoic acid, 98%, Thermo Scientific Chemicals
2-Amino-3-methoxybenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-3-methoxybenzoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 437720050
Price (USD)/ Each
115.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
136.00 
Save 20.35 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
115.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
136.00 
Save 20.35 (15%)
Add to cart
2-Amino-3-methoxybenzoic acid, 98%, Thermo Scientific Chemicals
Catalog number437720050
Price (USD)/ Each
115.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
136.00 
Save 20.35 (15%)
-
Add to cart
Chemical Identifiers
CAS3177-80-8
IUPAC Name2-amino-3-methoxybenzoic acid
Molecular FormulaC8H9NO3
InChI KeySXOPCLUOUFQBJV-UHFFFAOYSA-N
SMILESCOC1=CC=CC(C(O)=O)=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=97.5 %
Appearance (Color)White to yellow to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only