4-Cyanophenylacetic acid, 97%, Thermo Scientific Chemicals
4-Cyanophenylacetic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Cyanophenylacetic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 437730010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5462-71-5
IUPAC Name2-(4-cyanophenyl)acetic acid
Molecular FormulaC9H7NO2
InChI KeyWEBXRQONNWEETE-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
Melting point150.0°C to 155.0°C
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only