1x1 image pixel for data collection
TaqMan™ Array Human Inflammation Panel
Not what you're looking for? Search all of our TaqMan® arrays for your gene or pathway of interest.
Do you need this panel in a 384-well plate? Please visit Specialty Plating to learn more.
TaqMan™ Array Human Inflammation Panel
Applied Biosystems™

TaqMan™ Array Human Inflammation Panel

The Applied Biosystems™ TaqMan™ Human Inflammation Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis ofRead more
Have Questions?
Catalog NumberQuantity
43787074 cards
Catalog number 4378707
Price (USD)
2,505.00
Each
Add to cart
Quantity:
4 cards
Price (USD)
2,505.00
Each
Add to cart
The Applied Biosystems™ TaqMan™ Human Inflammation Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of inflammation genes important in drug discovery. This product is designed with 96 TaqMan™ Gene Expression Assays arrayed in 384 wells and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System and is sold in packs of 4.

The TaqMan™ Gene Signature Arrays for human inflammation contain over 90 genes that are, or have been, studied as targets for a range of inflammatory diseases. The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Format384-well microfluidic card
Internal Probe ModificationMGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
No. of Reactions1 x 2 μL Reactions
Product LineTaqMan™
Quantity4 cards
Research CategorySignal Transduction, Oncology, Biomarker-Related Pathway(s), Extracellular Matrix & Cell Adhesion, Inflammation
SpeciesHuman
TargetOver 90 genes that are targets for a range of inflammatory diseases
For Use With (Application)Gene Expression
FormDried Down
Label or DyeFAM
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored at 4°C.

Frequently asked questions (FAQs)

What are the storage recommendations for TaqMan Array Cards (TAC)?

We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection