2-Amino-3-bromo-5-chloropyridine, 97%, Thermo Scientific Chemicals
2-Amino-3-bromo-5-chloropyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-3-bromo-5-chloropyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number 437910050
Price (USD)/ Each
142.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
142.00
Add to cart
2-Amino-3-bromo-5-chloropyridine, 97%, Thermo Scientific Chemicals
Catalog number437910050
Price (USD)/ Each
142.00
-
Add to cart
Chemical Identifiers
CAS26163-03-1
IUPAC Name3-bromo-5-chloropyridin-2-amine
Molecular FormulaC5H4BrClN2
InChI KeyUWGGGYYCKDCTGN-UHFFFAOYSA-N
SMILESNC1=NC=C(Cl)C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point83°C to 87°C
Appearance (Color)White to light brown
GC>=96.0 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only