Thermo Scientific Chemicals

5-Amino-2-methyl-1,3-benzoxazole, 97%, Thermo Scientific Chemicals

Catalog number: 437920050
5 g, Each
Thermo Scientific Chemicals

5-Amino-2-methyl-1,3-benzoxazole, 97%, Thermo Scientific Chemicals

Catalog number: 437920050
5 g, Each
Quantity
Catalog number: 437920050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
72745-76-7
IUPAC Name
2-methyl-1,3-benzoxazol-5-amine
Molecular Formula
C8H8N2O
InChI Key
FECYTFWPNCBIHC-UHFFFAOYSA-N
SMILES
CC1=NC2=CC(N)=CC=C2O1
Appearance (Color)
Off-white to brown
Appearance (Form)
Crystalline powder or solid
Infrared spectrum
Conforms
Melting point
70°C to 76°C
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text