MicroAmp™ 96-Well Support Base
Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
MicroAmp™ 96-Well Support Base
Applied Biosystems™

MicroAmp™ 96-Well Support Base

Applied Biosystems™ MicroAmp™ 96-Well Support Bases are designed to hold MicroAmp™ Reaction Tubes in Tray/Retainer Sets or MicroAmp™ 48- andRead more
Have Questions?
Catalog NumberQuantity
437959010 bases
Catalog number 4379590
Price (USD)
119.00
Each
Add to cart
Quantity:
10 bases
Price (USD)
119.00
Each
Add to cart
Applied Biosystems™ MicroAmp™ 96-Well Support Bases are designed to hold MicroAmp™ Reaction Tubes in Tray/Retainer Sets or MicroAmp™ 48- and 96-Well Plates for convenient use in pre- and post-PCR sample processing.

• Convenient for pre- and post-PCR sample processing
• Easy to use for pipetting steps
• Designed to support 48-well and 96-well plate formats
Specifications
Certifications/ComplianceCertified DNA/RNase Free
For Use With (Application)PCR
For Use With (Equipment)2720 Thermal Cycler, 3500xL Dx Genetic Analyzer, 7000 System, 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, AB 2720 Thermal Cycler, GeneAmp 9700, QuantStudio™, SimpliAmp™ Thermal Cycler, StepOnePlus™, Veriti Dx Fast Thermal Cycler, Veriti Dx Thermal Cycler, Veriti Fast Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA™ 7 System, 3500xL Genetic Analyzer, QuantStudio™ Dx, ProFlex™ PCR System, ViiA™ 7 Dx Fast System, ViiA™ 7 Fast System, 3730xl DNA Analyzer, 7500 Fast Dx System, 3130 Genetic Analyzer, 2720 Thermal Cycler, 3730 DNA Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer
Packaging10 Bases
Plate Compatibility96-Well Plates
Product Type96-Well Plate Base
Quantity10 bases
Shipping ConditionRoom Temperature
ColorBlack
No. of Wells96
Product LineMicroAmp
Unit SizeEach
Contents & Storage
Contains 10 MicroAmp™ 96-Well Support Bases. Store at room temperature.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link