Informational:Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
Applied Biosystems™

MicroAmp™ 96-Well Tray for VeriFlex™ Blocks

Catalog number: 4379983
Have Questions?
Applied Biosystems™

MicroAmp™ 96-Well Tray for VeriFlex™ Blocks

Catalog number: 4379983
Have Questions?
Catalog Number
4379983
Unit Size
10 trays
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
437998310 trays
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Applied Biosystems™ MicroAmp™ 96-Well Tray for VeriFlex™ Blocks is specifically designed to snugly hold 0.1 mL MicroAmp™ Reaction Tubes with Caps, MicroAmp™ Fast Reaction Tubes with Caps, and MicroAmp™ Fast 8-Tube Strips.

• Engineered for efficient sample processing
• Designed specifically for 0.1 mL MicroAmp™ reaction tubes with caps
• Manufactured to high specification, ensuring a quality product

Convenience and Performance
The MicroAmp™ 96-Well Tray for VeriFlex™ Blocks allows you to organize your tubes while adding sample and mix.This handy tray reduces spillage and tube mix-ups by securing and ordering your tubes on the lab bench. When the samples are ready, simply put the tray of tubes directly on the block of the StepOne™ Real-Time PCR System or Veriti™ Thermal Cycler. Not only does the tray eliminate the hassle of transferring the tubes, but it also provides the thermal contact necessary for thermal cycling.

Specifications

Certifications/Compliance
Certified DNA/RNase Free
Product Line
MicroAmp
Color
Black
Product Type
Tray
For Use With (Equipment)
2720 Thermal Cycler, SimpliAmp™ Thermal Cycler, StepOnePlus™, Veriti Dx Fast Thermal Cycler, Veriti Dx Thermal Cycler, Veriti Fast Thermal Cycler, Veriti Thermal Cycler, ProFlex™ PCR System
For Use With (Application)
PCR
Plate Compatibility
96-Well Plates
Quantity
10 Trays
Shipping Condition
Room Temperature

Contents & Storage

Contains 10 MicroAmp™ 96-Well Trays for VeriFlex™ Blocks. Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text