tert-Butyl 2-amino-6,7-dihydro[1,3]thiazolo[5,4-c]pyridine-5(4H)-carboxylate, 97%, Thermo Scientific Chemicals
tert-Butyl 2-amino-6,7-dihydro[1,3]thiazolo[5,4-c]pyridine-5(4H)-carboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Butyl 2-amino-6,7-dihydro[1,3]thiazolo[5,4-c]pyridine-5(4H)-carboxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 438020010
Price (USD)/ Each
127.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
150.00 
Save 22.35 (15%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
127.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
150.00 
Save 22.35 (15%)
Add to cart
tert-Butyl 2-amino-6,7-dihydro[1,3]thiazolo[5,4-c]pyridine-5(4H)-carboxylate, 97%, Thermo Scientific Chemicals
Catalog number438020010
Price (USD)/ Each
127.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
150.00 
Save 22.35 (15%)
-
Add to cart
Chemical Identifiers
CAS365996-05-0
IUPAC Nametert-butyl 2-amino-4H,5H,6H,7H-[1,3]thiazolo[5,4-c]pyridine-5-carboxylate
Molecular FormulaC11H17N3O2S
InChI KeyBMLHPGOMLGKYIJ-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1CCC2=C(C1)SC(N)=N2
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)Off-white to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only