5-Amino-2,3-dihydrobenzofuran, 97%, Thermo Scientific Chemicals
5-Amino-2,3-dihydrobenzofuran, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Amino-2,3-dihydrobenzofuran, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 438030010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS42933-43-7
IUPAC Name2,3-dihydro-1-benzofuran-5-amine
Molecular FormulaC8H9NO
InChI KeyYJMADHMYUJFMQE-UHFFFAOYSA-N
SMILESNC1=CC2=C(OCC2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point80°C to 84°C
Infrared spectrumConforms
Appearance (Color)White to light brown
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only