Thermo Scientific Chemicals

Sodium hydrogencyanamide, 98%, Thermo Scientific Chemicals

Catalog number: 438050250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Sodium hydrogencyanamide, 98%, Thermo Scientific Chemicals

Catalog number: 438050250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 438050250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
17292-62-5
IUPAC Name
sodium cyanoazanide
Molecular Formula
CHN2Na
InChI Key
MBEGFNBBAVRKLK-UHFFFAOYSA-N
SMILES
[Na+].[NH-]C#N
Appearance (Color)
White to tan
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
Titration with HClO4
97.5 to 102.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text