Thermo Scientific Chemicals

Ethyl alpha-bromophenylacetate, 97%, Thermo Scientific Chemicals

Catalog number: 438060050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl alpha-bromophenylacetate, 97%, Thermo Scientific Chemicals

Catalog number: 438060050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 438060050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2882-19-1
IUPAC Name
ethyl 2-bromo-2-phenylacetate
Molecular Formula
C10H11BrO2
InChI Key
BKTKLDMYHTUESO-UHFFFAOYNA-N
SMILES
CCOC(=O)C(Br)C1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow
Refractive index
1.5370 to 1.5420 (20°C, 589 nm)
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text