Thermo Scientific Chemicals

Methyl picolinate, 98%, Thermo Scientific Chemicals

Catalog number: 438140250
25 g, Each
Thermo Scientific Chemicals

Methyl picolinate, 98%, Thermo Scientific Chemicals

Catalog number: 438140250
25 g, Each
Quantity
Catalog number: 438140250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2459-07-6
IUPAC Name
methyl pyridine-2-carboxylate
Molecular Formula
C7H7NO2
InChI Key
NMMIHXMBOZYNET-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC=N1
Appearance (Color)
Clear colorless to yellow to orange
GC
>=97.5 %
Infrared spectrum
Conforms
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text