5-Acetyl-2-methoxypyridine, 97%, Thermo Scientific Chemicals
5-Acetyl-2-methoxypyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Acetyl-2-methoxypyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 438170010
Price (USD)/ Each
64.80
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
64.80
Add to cart
5-Acetyl-2-methoxypyridine, 97%, Thermo Scientific Chemicals
Catalog number438170010
Price (USD)/ Each
64.80
-
Add to cart
Chemical Identifiers
CAS213193-32-9
IUPAC Name1-(6-methoxypyridin-3-yl)ethan-1-one
Molecular FormulaC8H9NO2
InChI KeyRYOQZXOVBJIUSX-UHFFFAOYSA-N
SMILESCOC1=NC=C(C=C1)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Melting point52.0°C to 57.0°C
GC>=96.0 %
Water=<3.00 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only