Informational:Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
Applied Biosystems™

MicroAmp™ 96-Well Tray/Retainer Set for Veriti™ Systems

Catalog number: 4381850
Have Questions?
Applied Biosystems™

MicroAmp™ 96-Well Tray/Retainer Set for Veriti™ Systems

Catalog number: 4381850
Have Questions?
Catalog Number
4381850
Unit Size
10 trays
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
438185010 trays
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Applied Biosystems™ MicroAmp™ 96-Well Tray/Retainer Set for Veriti™ Systems is specifically designed to snugly hold MicroAmp™ 8-Tube Strips (0.2 mL) and MicroAmp™ Reaction Tubes without Caps (0.2 mL) in a 96-well format for processing on the Veriti™ 96-Well Thermal Cycler.

• Engineered for efficient sample processing
• Designed specifically for MicroAmp™ 8-Tubes Strip (0.2 mL) and MicroAmp™ Reaction Tubes without Caps (0.2 mL)
• Manufactured to high specification, ensuring a quality product

Convenience and Performance The MicroAmp™ 96-Well Tray/Retainer Set for Veriti™ Systems allows you to organize your tubes while adding sample and mix. This handy tray reduces spillage and tube mix-ups by securing and ordering your tubes on the lab bench. When the samples are ready, simply put the tray of tubes directly on the block of the Veriti™ 96-Well Thermal Cycler. Not only does the tray eliminate the hassle of transferring the tubes, but it also provides the thermal contact necessary for thermal cycling.

Note: See user's manual or package insert for limited label license and trademark information.

Specifications

Certifications/Compliance
Certified DNA/RNase Free
Product Line
MicroAmp
Color
Blue
Product Type
Tray/Retainer Set
For Use With (Equipment)
SimpliAmp™ Thermal Cycler, Veriti Dx Thermal Cycler, Veriti Thermal Cycler, ProFlex™ PCR System
For Use With (Application)
PCR
Plate Compatibility
96-Well Plates
Quantity
10 Trays
Shipping Condition
Room Temperature

Contents & Storage

Contains 10 MicroAmp™ 96-Well Tray⁄Retainer Sets. Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text