1x1 image pixel for data collection
Peak Scanner™ Software v1.0
Access the new Peak Scanner application on Thermo Fisher Cloud here »
Product Image
Applied Biosystems™
Peak Scanner™ Software v1.0
Peak Scanner Software v1.0 is a DNA sizing software that can either be downloaded for free or purchased for freeRead more
Have Questions?
Catalog NumberQuantity
43818671 software
Catalog number 4381867
Price (USD)
-
Quantity:
1 software
Peak Scanner Software v1.0 is a DNA sizing software that can either be downloaded for free or purchased for free as a software kit.
Use this software to perform DNA fragment analysis, separate a mixture of DNA fragments according to their sizes, provide a profile of the separation, and precisely calculate the sizes of the fragments. The software allows you to view, edit, analyze, print, and export fragment analysis data generated using the Applied Biosystems™ genetic analyzers.
•Use a workflow panel to guide you through analysis
•Quickly analyze samples with the default sizing methods
•View or print thumbnail and checkerboard view
•View raw and analyzed data simultaneously
•Analyze large fragments (e.g., 1200 bp)
•Get better results with linear and non-linear sizing algorithms
Easy to use
Peak Scanner™ Software v1.0 guides you through the process of fragment analysis. If you just want to quickly perform an analysis, you can use default analysis methods to get the answers you need. Having problems? Use the latest troubleshooting tools. The clear iconography and simple project archiving means this tool is the easiest way to perform DNA fragment analysis.
Customizable
Customize Peak Scanner™ Software v1.0 to streamline your process. Adjust thumbnail and checkerboard viewing and printing options, change the sample and sizing tables, and edit locking and selective analysis, including peak labeling, merging and splitting. You can even overlay sizing curves on analyzed data.
Flexible
Peak Scanner™ Software v1.0 enables the simultaneous viewing of raw and analyzed data. Now you have the ability to size large fragments such as 1200 bp sized fragments. Use either linear or non-linear sizing algorithms. This flexible software tool has the potential to be used in many novel applications.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Data EntryGenetic Analyzer data (.fsa format)
Data OutputsElectropherogram Analysis Reports
For Use With (Equipment)3100 Genetic Analyzer, 3730 DNA Analyzer
LicenseStatic
Operating SystemWindows XP (Service Pack 2), Windows 2000 (Service Pack 4)
Product TypePeak Scanner Software V1.0
Quantity1 software
Software CategorySoftware
Product LinePeak Scanner™
Software TypeFragment Sizing
Starting MaterialDNA
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link