Methyl tridecanoate, 97%, Thermo Scientific Chemicals
Methyl tridecanoate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl tridecanoate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
10 g
Catalog number 438200010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1731-88-0
IUPAC Namemethyl tridecanoate
Molecular FormulaC14H28O2
InChI KeyJNDDPBOKWCBQSM-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only