Thermo Scientific Chemicals

3,5-Di-tert-butylbenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 438220050
5 g, Each
Thermo Scientific Chemicals

3,5-Di-tert-butylbenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 438220050
5 g, Each
Quantity
Catalog number: 438220050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
16225-26-6
IUPAC Name
3,5-di-tert-butylbenzoic acid
Molecular Formula
C15H22O2
InChI Key
NCTSLPBQVXUAHR-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC(=CC(=C1)C(O)=O)C(C)(C)C
Appearance (Color)
White to off-white
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
170°C to 175°C
HPLC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text