Thermo Scientific Chemicals

tert-Butyl 4-(2-chloroethyl)piperazine-1-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: 438240050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

tert-Butyl 4-(2-chloroethyl)piperazine-1-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: 438240050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 438240050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
208167-83-3
IUPAC Name
tert-butyl 4-(2-chloroethyl)piperazine-1-carboxylate
Molecular Formula
C11H21ClN2O2
InChI Key
MYOWELLYEZMECA-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)N1CCN(CCCl)CC1
Melting point
62°C to 68°C
GC
>=96.0 %
Appearance (Color)
White to light brown
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text