2-Methyl-2-phenylpropanenitrile, 97%, Thermo Scientific Chemicals
2-Methyl-2-phenylpropanenitrile, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methyl-2-phenylpropanenitrile, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 438250010
Price (USD)/ Each
90.70
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
90.70
Add to cart
2-Methyl-2-phenylpropanenitrile, 97%, Thermo Scientific Chemicals
Catalog number438250010
Price (USD)/ Each
90.70
-
Add to cart
Chemical Identifiers
CAS1195-98-8
IUPAC Name2-methyl-2-phenylpropanenitrile
Molecular FormulaC10H11N
InChI KeyPGQTYXFMSZUGOW-UHFFFAOYSA-N
SMILESCC(C)(C#N)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only