Thermo Scientific Chemicals

Isobutanesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 438410050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Isobutanesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 438410050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 438410050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
35432-36-1
IUPAC Name
2-methylpropane-1-sulfonyl chloride
Molecular Formula
C4H9ClO2S
InChI Key
NQLNHTOCFWUYQE-UHFFFAOYSA-N
SMILES
CC(C)CS(Cl)(=O)=O
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to brown
Appearance (Form)
Liquid
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text