N-BOC-Pyrrole, 98%, Thermo Scientific Chemicals
N-BOC-Pyrrole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-BOC-Pyrrole, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 438511000
Price (USD)/ Each
201.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
237.00 
Save 35.35 (15%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
201.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
237.00 
Save 35.35 (15%)
Add to cart
N-BOC-Pyrrole, 98%, Thermo Scientific Chemicals
Catalog number438511000
Price (USD)/ Each
201.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
237.00 
Save 35.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nametert-butyl 1H-pyrrole-1-carboxylate
Molecular FormulaC9H13NO2
InChI KeyIZPYBIJFRFWRPR-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1C=CC=C1
Molecular Weight (g/mol)167.21
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only