Strontium nitrate, 98%, extra pure, Thermo Scientific Chemicals
Strontium nitrate, 98%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Strontium nitrate, 98%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Packaging:
Plastic bottle
Catalog number 438520050
Price (USD)/ Each
292.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
344.00 
Save 51.35 (15%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
292.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
344.00 
Save 51.35 (15%)
Add to cart
Strontium nitrate, 98%, extra pure, Thermo Scientific Chemicals
Catalog number438520050
Price (USD)/ Each
292.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
344.00 
Save 51.35 (15%)
-
Add to cart
Chemical Identifiers
CAS10042-76-9
IUPAC Namestrontium(2+) dinitrate
Molecular FormulaN2O6Sr
InChI KeyDHEQXMRUPNDRPG-UHFFFAOYSA-N
SMILES[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric>=97.5 %
Heavy metals (as Pb)=<10 ppm
Loss on drying=<0.5 %
pH5 to 7 (5 % at 20°C)
Appearance (Form)Crystalline powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only