Thermo Scientific Chemicals

3-Ethynylthiophene, 96%, Thermo Scientific Chemicals

Catalog number: 438580050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Ethynylthiophene, 96%, Thermo Scientific Chemicals

Catalog number: 438580050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 438580050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
67237-53-0
IUPAC Name
3-ethynylthiophene
Molecular Formula
C6H4S
InChI Key
MJHLPKWONJUCFK-UHFFFAOYSA-N
SMILES
C#CC1=CSC=C1
Appearance
May darken on storage
Infrared spectrum
Conforms
GC
>=95.0 %
Appearance (Color)
Colorless to yellow to orange
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text