Methyl benzenesulfinate, 98%, Thermo Scientific Chemicals
Methyl benzenesulfinate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl benzenesulfinate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Catalog number 438610250
Price (USD)/ Each
-
Quantity:
25 mL
Request bulk or custom format
Chemical Identifiers
CAS670-98-4
IUPAC Namemethyl benzenesulfinate
Molecular FormulaC7H8O2S
InChI KeyPSNSVDSRLUYDKF-UHFFFAOYNA-N
SMILESCOS(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)Colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only