N-Hydroxysulfosuccinimide sodium salt, 95%, Thermo Scientific Chemicals
N-Hydroxysulfosuccinimide sodium salt, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Hydroxysulfosuccinimide sodium salt, 95%, Thermo Scientific Chemicals

Coupling reagent
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 438650010
Price (USD)/ Each
272.65
Online exclusive
320.00 
Save 47.35 (15%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
272.65
Online exclusive
320.00 
Save 47.35 (15%)
Add to cart
N-Hydroxysulfosuccinimide sodium salt, 95%, Thermo Scientific Chemicals
Catalog number438650010
Price (USD)/ Each
272.65
Online exclusive
320.00 
Save 47.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namesodium 1-hydroxy-2,5-dioxopyrrolidine-3-sulfonate
Molecular FormulaC4H4NNaO6S
InChI KeyRPENMORRBUTCPR-UHFFFAOYNA-M
SMILES[Na+].ON1C(=O)CC(C1=O)S([O-])(=O)=O
Molecular Weight (g/mol)217.13
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only