Thermo Scientific Chemicals

FMOC isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog number: 438660050
5 g, Each
Thermo Scientific Chemicals

FMOC isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog number: 438660050
5 g, Each
Quantity
Catalog number: 438660050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
199915-38-3
IUPAC Name
(9H-fluoren-9-yl)methyl N-carbothioylcarbamate
Molecular Formula
C16H11NO2S
InChI Key
DHMYULZVFHHEHE-UHFFFAOYSA-N
SMILES
O=C(OCC1C2=CC=CC=C2C2=CC=CC=C12)N=C=S
Infrared spectrum
Conforms
Appearance (Form)
Low melting solid
Elemental Analysis
4.8 to 5.2 % (N)
Appearance (Color)
White to yellow
Elemental Analysis
65.6 to 71.0 % (C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text