Thermo Scientific Chemicals

(3-Bromopropoxy)-tert-butyldimethylsilane, 97%, stabilized over sodium carbonate, Thermo Scientific Chemicals

Catalog number: 438690250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

(3-Bromopropoxy)-tert-butyldimethylsilane, 97%, stabilized over sodium carbonate, Thermo Scientific Chemicals

Catalog number: 438690250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 438690250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
89031-84-5
IUPAC Name
(3-bromopropoxy)(tert-butyl)dimethylsilane
Molecular Formula
C9H21BrOSi
InChI Key
QGMROEZDWJTIDW-UHFFFAOYSA-N
SMILES
CC(C)(C)[Si](C)(C)OCCCBr
GC
>=96.0 %
Appearance (Color)
Colorless
Infrared spectrum
Conforms
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text